Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRK7
Entrez Gene ID
714916
Gene Name
G protein-coupled receptor kinase 7
Gene Aliases
-
Chromosome Location
Chr.2: 43907434 - 43948075 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001112570.4 XP_001112570.2 - 2136 109

View Full Product Details