Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYBA1
Entrez Gene ID
716819
Gene Name
crystallin beta A1
Gene Aliases
EGK_08349
Chromosome Location
Chr.16: 24882830 - 24890358 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194386.1 NP_001181315.1 - 363 116
XM_028835737.1 XP_028691570.1 - 862 116

View Full Product Details