Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PROCR
Entrez Gene ID
706040
Gene Name
protein C receptor
Gene Aliases
EGK_02471
Chromosome Location
Chr.10: 70377078 - 70423582 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015149144.2 XP_015004630.1 - 542 143
XM_028828348.1 XP_028684181.1 - 540 143

View Full Product Details