Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL17RB
Entrez Gene ID
693671
Gene Name
interleukin 17 receptor B
Gene Aliases
EGK_11555
Chromosome Location
Chr.2: 110036402 - 110057335 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082504.4 XP_001082504.2 - 634 90
XM_015130684.2 XP_014986170.1 - 337 90
GenBank mRNA JU335345.1 - - 518 90
JV586599.1 - - 119 90

View Full Product Details