Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRA10AC1
Entrez Gene ID
700389
Gene Name
FRA10A associated CGG repeat 1
Gene Aliases
C10orf4, C9H10orf4
Chromosome Location
Chr.9: 93829024 - 93863246 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266720.2 NP_001253649.1 - 357 175
XM_015147634.2 XP_015003120.1 - 409 175
XM_015147636.2 XP_015003122.1 - 708 175
XM_028853355.1 XP_028709188.1 - 318 175
XM_028853356.1 XP_028709189.1 - 409 175
GenBank mRNA JU322797.1 - - 461 175
JU472827.1 - - 450 175
JV045627.1 - - 458 175
JV565064.1 - - 357 175

View Full Product Details