Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC17
Entrez Gene ID
706591
Gene Name
coiled-coil domain containing 17
Gene Aliases
-
Chromosome Location
Chr.1: 178793960 - 178798715 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002801532.3 XP_002801578.2 - 2038 133

View Full Product Details