Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC1
Entrez Gene ID
696721
Gene Name
doublecortin domain containing 1
Gene Aliases
DCDC5
Chromosome Location
Chr.14: 34052049 - 34553887 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015114578.2 XP_014970064.2 - 989 95
XM_015114580.2 XP_014970066.2 - 989 95
XM_028833537.1 XP_028689370.1 - 989 95
XM_028833538.1 XP_028689371.1 - 989 95
XM_028833539.1 XP_028689372.1 - 989 95
XM_028833540.1 XP_028689373.1 - 989 95

View Full Product Details