Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HACD2
Entrez Gene ID
715374
Gene Name
3-hydroxyacyl-CoA dehydratase 2
Gene Aliases
PTPLB
Chromosome Location
Chr.2: 152727687 - 152826545 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261129.1 NP_001248058.1 - 699 155
XM_028843390.1 XP_028699223.1 - 503 155
XM_028843391.1 XP_028699224.1 - 746 155
XM_028843392.1 XP_028699225.1 - 409 155
GenBank mRNA JU327265.1 - - 674 155
JV044709.1 - - 699 155
JV639352.1 - - 695 155

View Full Product Details