Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFCAB12
Entrez Gene ID
703176
Gene Name
EF-hand calcium binding domain 12
Gene Aliases
-
Chromosome Location
Chr.2: 147068331 - 147095847 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001094977.4 XP_001094977.2 - 1615 167

View Full Product Details