Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMPK2
Entrez Gene ID
708602
Gene Name
cytidine/uridine monophosphate kinase 2
Gene Aliases
-
Chromosome Location
Chr.13: 101909431 - 101926277 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001099309.4 XP_001099309.1 - 878 101
GenBank mRNA JU338826.1 - - 150 101

View Full Product Details