Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GJC1
Entrez Gene ID
715463
Gene Name
gap junction protein gamma 1
Gene Aliases
CXNQ, EGK_08814
Chromosome Location
Chr.16: 56071022 - 56099249 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194656.1 NP_001181585.1 - 270 110
XM_028835702.1 XP_028691535.1 - 1293 110
XM_028835703.1 XP_028691536.1 - 305 110
GenBank mRNA JU473843.1 - - 81 110
JV043296.1 - - 174 110

View Full Product Details