Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WEE2
Entrez Gene ID
694392
Gene Name
WEE2 oocyte meiosis inhibiting kinase
Gene Aliases
WEE1B
Chromosome Location
Chr.3: 168103987 - 168125745 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105546.3 NP_001099016.3 - 1832 74

View Full Product Details