Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARMCX5
Entrez Gene ID
100427780
Gene Name
armadillo repeat containing X-linked 5
Gene Aliases
EGK_21134
Chromosome Location
Chr.X: 99225812 - 99230790 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194914.2 NP_001181843.1 - 2056 101
XM_015127844.2 XP_014983330.1 - 2362 101
XM_015127845.2 XP_014983331.1 - 2516 101
XM_015127846.2 XP_014983332.1 - 2471 101
GenBank mRNA JU323327.1 - - 2207 101
JU472361.1 - - 2291 101

View Full Product Details