Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD1A
Entrez Gene ID
698438
Gene Name
CD1a molecule
Gene Aliases
-
Chromosome Location
Chr.1: 92753887 - 92770766 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145818.1 NP_001139290.1 - 61 95
GenBank mRNA AB458511.1 - - 61 95

View Full Product Details