Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LORICRIN
Entrez Gene ID
714526
Gene Name
loricrin cornified envelope precursor protein
Gene Aliases
LOR
Chromosome Location
Chr.1: 97806861 - 97807874 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204365.2 NP_001191294.2 - 713 77

View Full Product Details