Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDKN1B
Entrez Gene ID
697188
Gene Name
cyclin dependent kinase inhibitor 1B
Gene Aliases
EGK_03400
Chromosome Location
Chr.11: 14091595 - 14096810 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261005.1 NP_001247934.1 - 957 78
GenBank mRNA JU321114.1 - - 915 78
JU472875.1 - - 952 78
JV044784.1 - - 957 78
JV529662.1 - - 967 78

View Full Product Details