Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH4
Entrez Gene ID
714347
Gene Name
junctophilin 4
Gene Aliases
-
Chromosome Location
Chr.7: 85608670 - 85619478 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261095.1 NP_001248024.1 - 1948 71
XM_015143320.2 XP_014998806.1 - 2480 71
GenBank mRNA JU336742.1 - - 2074 71
JU336743.1 - - 1948 71
JV560420.1 - - 2056 71

View Full Product Details