Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klf6
Entrez Gene ID
58954
Gene Name
Kruppel-like factor 6
Gene Aliases
Aa1017, CPBP, Copeb, Zf9
Chromosome Location
Chr.17: 67887939 - 67945038 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031642.4 NP_113830.2 3 - 4 1044 65
GenBank mRNA BC097306.1 - 3 - 4 969 65
FQ225029.1 - 2 - 3 600 65
FQ231202.1 - 2 - 3 384 65
FQ231451.1 - 2 - 3 384 65
FQ232231.1 - 2 - 3 383 65

View Full Product Details