The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012598.2 | NP_036730.1 | 2 - 3 | 438 | 73 |
GenBank mRNA | BC081836.1 | - | 2 - 3 | 438 | 73 |
L03294.1 | - | - | 423 | 73 |