The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270869.1 | NP_001257798.1 | 1 - 2 | 217 | 105 |
NM_031073.3 | NP_112335.3 | 2 - 3 | 299 | 105 | |
GenBank mRNA | BC070504.1 | - | 2 - 3 | 299 | 105 |
FM101680.1 | - | 1 - 2 | 216 | 105 | |
M34643.1 | - | 1 - 2 | 50 | 105 |