The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031126.1 | NP_112388.1 | 14 - 15 | 1252 | 63 |
XM_006248783.2 | XP_006248845.1 | 14 - 15 | 1299 | 63 | |
GenBank mRNA | BC097310.1 | - | 13 - 14 | 1864 | 63 |
U20283.1 | - | 14 - 15 | 1252 | 63 |