The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031732.2 | NP_113920.2 | 5 - 6 | 645 | 85 |
GenBank mRNA | BC088125.1 | - | 5 - 6 | 645 | 85 |
FQ219411.1 | - | 5 - 6 | 675 | 85 | |
L22339.1 | - | 4 - 5 | 555 | 85 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006244237.2 | XP_006244299.1 | - | 562 | 85 |