The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053548.1 | NP_446000.1 | 4 - 5 | 730 | 79 |
GenBank mRNA | AF031528.1 | - | 3 - 4 | 604 | 79 |