The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144739.1 | NP_653340.1 | 5 - 6 | 977 | 85 |
GenBank mRNA | AF458413.1 | - | 5 - 6 | 977 | 85 |
BC070883.1 | - | 5 - 6 | 705 | 85 |