Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Camk2b
Entrez Gene ID
24245
Gene Name
calcium/calmodulin-dependent protein kinase II beta
Gene Aliases
Ck2b
Chromosome Location
Chr.14: 86208876 - 86297751 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042354.1 NP_001035813.1 10 - 11 903 69
NM_001042356.1 NP_001035815.1 10 - 11 903 69
NM_021739.2 NP_068507.2 10 - 11 903 69
XM_006251170.2 XP_006251232.1 10 - 11 1002 69
XM_006251171.2 XP_006251233.1 10 - 11 1002 69
XM_006251172.2 XP_006251234.1 10 - 11 1002 69
XM_006251174.2 XP_006251236.1 10 - 11 1002 69
XM_006251176.2 XP_006251238.1 10 - 11 983 69
XM_006251177.2 XP_006251239.1 10 - 11 982 69
GenBank mRNA M16112.1 - 10 - 11 883 69
X83375.1 - 10 - 11 865 69

View Full Product Details