Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcer2
Entrez Gene ID
171075
Gene Name
Fc fragment of IgE, low affinity II, receptor for (CD23)
Gene Aliases
CD23, Fcer2a
Chromosome Location
Chr.12: 2233772 - 2245324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033924.1 NP_001029096.1 10 - 11 802 78
NM_133550.2 NP_598234.2 9 - 10 739 78
XM_008768939.1 XP_008767161.1 9 - 10 966 78
GenBank mRNA AF381978.1 - 9 - 10 688 78
BC078749.1 - 10 - 11 802 78
X73579.1 - 8 - 9 622 78

View Full Product Details