Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alg10
Entrez Gene ID
245960
Gene Name
ALG10, alpha-1,2-glucosyltransferase
Gene Aliases
Alg10b, KCR1
Chromosome Location
Chr.7: 131330913 - 131336183 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139101.2 NP_620801.1 1 - 2 270 64
GenBank mRNA U78090.1 - - 270 64

View Full Product Details