The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019182.2 | NP_062055.1 | 7 - 8 | 710 | 75 |
XM_008770271.1 | XP_008768493.1 | 7 - 8 | 742 | 75 | |
XM_008770272.1 | XP_008768494.1 | 7 - 8 | 697 | 75 | |
XM_008770273.1 | XP_008768495.1 | 7 - 8 | 830 | 75 | |
GenBank mRNA | AF022081.1 | - | 7 - 8 | 628 | 75 |
AY050655.1 | - | 7 - 8 | 722 | 75 | |
BC062024.1 | - | 7 - 8 | 710 | 75 | |
FQ211855.1 | - | 6 - 7 | 592 | 75 |