Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Steap3
Entrez Gene ID
170824
Gene Name
STEAP family member 3, metalloreductase
Gene Aliases
-
Chromosome Location
Chr.13: 36257220 - 36274144 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133314.1 NP_579848.1 1 - 2 520 97
GenBank mRNA AF238865.1 - 1 - 2 519 97
AF335281.1 - 1 - 2 520 97
BC085696.1 - - 541 97

View Full Product Details