The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024290.1 | NP_001019461.1 | 2 - 3 | 381 | 69 |
GenBank mRNA | BC094516.1 | - | 2 - 3 | 381 | 69 |