Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bpifb6
Entrez Gene ID
311558
Gene Name
BPI fold containing family B, member 6
Gene Aliases
Bpil3, Lplunc6
Chromosome Location
Chr.3: 149384571 - 149396130 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107791.1 NP_001101261.1 4 - 5 273 66
XM_006235317.1 XP_006235379.1 13 - 14 1233 66

View Full Product Details