Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pax3
Entrez Gene ID
114502
Gene Name
paired box 3
Gene Aliases
-
Chromosome Location
Chr.9: 84004004 - 84101230 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053710.1 NP_446162.1 8 - 9 1803 85
XM_006245131.2 XP_006245193.1 8 - 9 1857 85
XM_008767181.1 XP_008765403.1 9 - 10 1936 85

View Full Product Details