The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109007.2 | NP_001102477.2 | 21 - 22 | 2681 | 62 |
NM_001172159.1 | NP_001165630.1 | 20 - 21 | 2639 | 62 |