The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108470.1 | NP_001101940.1 | 4 - 5 | 1252 | 67 |
XM_006228116.2 | XP_006228178.1 | 5 - 6 | 1139 | 67 | |
XM_008758767.1 | XP_008756989.1 | 5 - 6 | 1400 | 67 | |
XM_008758768.1 | XP_008756990.1 | 4 - 5 | 1507 | 67 |