Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plac9
Entrez Gene ID
361105
Gene Name
placenta-specific 9
Gene Aliases
-
Chromosome Location
Chr.16: 3851102 - 3866008 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108395.1 NP_001101865.1 1 - 2 70 94
XM_006252630.2 XP_006252692.1 1 - 2 319 94

View Full Product Details