Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcer2
Entrez Gene ID
171075
Gene Name
Fc fragment of IgE, low affinity II, receptor for (CD23)
Gene Aliases
CD23, Fcer2a
Chromosome Location
Chr.12: 2233772 - 2245324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033924.1 NP_001029096.1 2 - 3 138 69
NM_133550.2 NP_598234.2 2 - 3 138 69
GenBank mRNA BC078749.1 - 2 - 3 138 69

View Full Product Details