Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex11g
Entrez Gene ID
288369
Gene Name
peroxisomal biogenesis factor 11 gamma
Gene Aliases
Pex11c
Chromosome Location
Chr.12: 2000426 - 2007516 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105902.1 NP_001099372.1 1 - 2 112 65

View Full Product Details