Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam53c
Entrez Gene ID
685405
Gene Name
family with sequence similarity 53, member C
Gene Aliases
-
Chromosome Location
Chr.18: 27558091 - 27572633 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772055.1 XP_008770277.1 2 - 3 310 88

View Full Product Details