The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191986.1 | NP_001178915.1 | - | 1246 | 95 |
XM_006241085.2 | XP_006241147.1 | - | 2398 | 95 | |
XM_006241094.2 | XP_006241156.1 | - | 315 | 95 |