Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gimap8
Entrez Gene ID
500112
Gene Name
GTPase, IMAP family member 8
Gene Aliases
IanT
Chromosome Location
Chr.4: 78283357 - 78294838 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033923.1 NP_001029095.1 1 - 2 246 99
XM_006236441.2 XP_006236503.1 2 - 3 223 99
GenBank mRNA AJ633685.1 - 1 - 2 87 99
BC099228.1 - 1 - 2 246 99
DQ125335.1 - 1 - 2 276 99
DQ125336.1 - 1 - 2 276 99

View Full Product Details