Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC679924
Entrez Gene ID
679924
Gene Name
similar to Kif19A CG9913-PB, isoform B
Gene Aliases
-
Chromosome Location
Chr.12: 16233512 - 16283177 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248973.2 XP_006249035.1 6 - 7 837 63

View Full Product Details