Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Parm1
Entrez Gene ID
286894
Gene Name
prostate androgen-regulated mucin-like protein 1
Gene Aliases
CIPAR-1, Cipar1, PARM-1
Chromosome Location
Chr.14: 17972914 - 18076870 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC060539.1 - 1 - 2 246 58
FQ219991.1 - 1 - 2 243 58

View Full Product Details