The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031022.1 | NP_112284.1 | 9 - 10 | 5209 | 159 |
GenBank mRNA | X56541.2 | - | 9 - 10 | 5209 | 159 |