The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008876.2 | NP_001008876.1 | 4 - 5 | 330 | 122 |
GenBank mRNA | AY310144.1 | - | 4 - 5 | 330 | 122 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008767502.1 | XP_008765724.1 | - | 903 | 122 |