Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfyve21
Entrez Gene ID
362789
Gene Name
zinc finger, FYVE domain containing 21
Gene Aliases
-
Chromosome Location
Chr.6: 136380934 - 136400810 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108725.1 NP_001102195.1 3 - 4 407 131
XM_006240639.2 XP_006240701.2 3 - 4 463 131
XM_006240641.2 XP_006240703.2 3 - 4 463 131
XM_006240642.2 XP_006240704.2 3 - 4 463 131
XM_006240643.2 XP_006240705.2 3 - 4 463 131
XM_006240644.2 XP_006240706.2 3 - 4 463 131
XM_008764946.1 XP_008763168.1 3 - 4 463 131

View Full Product Details