Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tecr
- Entrez Gene ID
-
191576
- Gene Name
- trans-2,3-enoyl-CoA reductase
- Gene Aliases
-
Gpsn2, SC2
- Chromosome Location
-
Chr.19: 24705478 - 24732029 on Build Rnor_6.0
- Gene Symbol
- RGD1560015
- Entrez Gene ID
-
499018
- Gene Name
- similar to glycoprotein, synaptic 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 47587022 - 47588091 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_574311.6 |
XP_574311.1 |
1 - 1 |
845 |
94 |
View Full Product Details