The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017024.2 | NP_058720.2 | 5 - 6 | 757 | 142 |
XM_006255441.1 | XP_006255503.1 | 5 - 6 | 1226 | 142 | |
GenBank mRNA | BC091155.1 | - | 5 - 6 | 757 | 142 |
FQ209624.1 | - | 5 - 6 | 772 | 142 | |
FQ218245.1 | - | 5 - 6 | 772 | 142 | |
FQ218883.1 | - | 5 - 6 | 775 | 142 | |
U62803.1 | - | 5 - 6 | 760 | 142 | |
X54096.1 | - | 5 - 6 | 771 | 142 |