Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tctex1d1
Entrez Gene ID
362553
Gene Name
Tctex1 domain containing 1
Gene Aliases
RGD1311932
Chromosome Location
Chr.5: 122498884 - 122522110 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135030.1 NP_001128502.1 2 - 3 321 95
XM_008763889.1 XP_008762111.1 2 - 3 2994 95

View Full Product Details