Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan15
Entrez Gene ID
679462
Gene Name
tetraspanin 15
Gene Aliases
-
Chromosome Location
Chr.20: 31812558 - 31857191 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115032.1 NP_001108504.1 4 - 5 549 135
GenBank mRNA BC158654.1 - 4 - 5 549 135

View Full Product Details