Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sdccag3
Entrez Gene ID
306322
Gene Name
serologically defined colon cancer antigen 3
Gene Aliases
-
Chromosome Location
Chr.3: 3827498 - 3834219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013135.2 NP_001013153.2 4 - 5 510 85
XM_006233681.2 XP_006233743.1 3 - 4 420 85
XM_006233682.2 XP_006233744.1 3 - 4 339 85
XM_006233683.2 XP_006233745.1 4 - 5 399 85
XM_006233684.2 XP_006233746.1 2 - 3 252 85
XM_006233685.2 XP_006233747.1 2 - 3 276 85
XM_006233686.2 XP_006233748.1 4 - 5 489 85
GenBank mRNA BC168170.1 - 2 - 3 282 85

Gene Symbol
LOC100363321
Entrez Gene ID
100363321
Gene Name
serologically defined colon cancer antigen 3
Gene Aliases
-
Chromosome Location
Chr.16: 18379595 - 18381631 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252792.1 XP_006252854.1 1 - 1 275 85

Gene Symbol
RGD1562289
Entrez Gene ID
501516
Gene Name
similar to RIKEN cDNA C330016H24
Gene Aliases
-
Chromosome Location
Chr.X: 7049662 - 7051625 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_576917.5 XP_576917.1 1 - 1 344 85

View Full Product Details